Wieder Tongue Depressor, OR Grade, Sklar
Fournisseur: Sklar
Certificats
A propos de cet article
Wieder tongue depressor is used to press down the tongue for a clear view during examination of the mouth and throat.
- Made from stainless steel
- Available in two sizes
- Curved solid handle
- Serrated surface
- Latex-free
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.