Spécifications
- Length:13.3 cm (51/4")
- Description:Tieck-Halle Infant Nasal Speculum
- Material:Stainless Steel
- Cat. no.:10198-512
- Supplier No.:70-1040
Spécifications
A propos de cet article
Sklar's Tieck-Halle Infant Nasal Speculum is used for retracting nasal walls for examination of the intranasal cavity.
- Double spring and non-ratcheted
- Plier handle instrument
This device has 16 mm jaws that are slightly angled so as not to obstruct the line of view of the surgeon.