Spécifications
- Description:Hartmann Ear Speculum
- Material:Stainless Steel
- Tip Configuration:Round
- Tip Dimensions:4 mm
- Cat. no.:10198-408
- Supplier No.:67-6004
Spécifications
A propos de cet article
The Sklar Hartman Ear Speculum is a cone/funnel-shaped attachment for an otoscope that allows examination of the ear canal and eardrum
The speculum is inserted into the ear canal and the otoscope handle provides a light source and low-power magnification so the practitioner can view the eardrum. Disease and infections of the middle or inner ear can largely be detected by this examination. This device is a round attachment with a 4mm diameter.