3B Scientific® Ring Mount Skeleton
# de catalogue 470029-944
Fournisseur: American 3B Scientific
Spécifications
- Description:Skeleton And Stand-Ring Mount
- Height:189 cm
- Cat. no.:470029-944
- Length:53 cm
- Width:53 cm
- Supplier No.:A10/1
Spécifications
A propos de cet article
An Economical Recreation of The Human Skeleton!
- Natural Skeleton Size
- Stable Metal Base with Casters
- Three-Year Manufacturers Guarantee
- Includes 3B Smart Anatomy
This model meets the requirements of introductory anatomy courses without breaking your budget. Features include detachable limbs and removable skull with a sectioned calvarium for up-close study of various skeletal parts. In addition, the jaw, with particularly fine dentition, is hinged to demonstrate mouth movement. Constructed with durable metal hardware, the ring-mounted skeleton, on a wheeled stand, is suspended from the fitting secured at the top of the skull. Includes a plastic dust cover.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought