ContactUsLinkComponent
Steiner Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Steiner Auvard Weighted Vaginal Speculum, OR Grade, Sklar
# de catalogue 10197-198
Fournisseur:  Sklar
Speculum
Steiner Auvard Weighted Vaginal Speculum, OR Grade, Sklar
# de catalogue 10197-198
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Sterility:
    Non sterile
  • Description:
    Steiner Auvard Weighted Vaginal Speculum, 5.5×1.25" Blades
  • Disposable / reusable:
    Reusable
  • Latex/Latex-free:
    Latex-free
  • Material:
    Stainless Steel
  • Curvature:
    Straight
  • Grade:
    Premium OR-Grade
  • Supplier No.:
    90-3559
Spécifications
A propos de cet article
The Sklar Steiner-Auvard Weighted Vaginal Speculum is a single end vaginal speculum commonly used in OB/GYN procedures, such as vaginal hysterectomy or dilation and curettage.
This speculum features extra long blades. It is weighted with a medium, 21/2 lbs. weight.
Customers Who Bought This Also Bought