Spécifications
- Description:Henrotin Weighted Vaginal Speculum
- Material:Stainless Steel
- Tip Configuration:31/4×11/4" Blade
- Curvature:Straight
- Cat. no.:10197-196
- Supplier No.:90-3555
Spécifications
A propos de cet article
Sklar's Henrotin Weighted Vaginal Speculum utilizes a 3 lbs weight to hold the narrow, angled blade in place for use in OB/GYN procedures. The blade is angle downward 30® to provide the user with a greater view of the vaginal canal.