FlexTech™ Seamless Knit Palm Coated Glove, Sandy Nitrile Palm, Wells Lamont®
Fournisseur: Wells Lamont
Certificats
A propos de cet article
The seamless, machine-knit shell of these gloves provides all-day comfort and protection.
- Sandy nitrile palm coating for grip in wet and oily conditions
- Washing machine friendly
- Cut resistant
- Applications in metal handling, metal stamping, and automotive manufacturing
Certifications: ASTM F1790-97. ANSI Cut 5.
Ordering information: Gloves are sold by the dozen pairs.
Spécifications
- Catalog No:
- 10847-278
- 10847-276
- 10847-302
- 10847-300
- 10847-298
- 10847-296
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free