To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Actin Alpha 1, Skeletal Muscle
- Clonality:Polyclonal
- Gene ID:58
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:ACTA1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Size:400 µL
- Western blot:Yes
- Epitope:C-Terminal
- Antigen synonyms:ACT1|alpha skeletal muscle Actin|ASMA|alpha Actin 1|NEM2|ACTA|NEM1
- Amino acid number:346 to 375
- Storage buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular weight:42.051 kDa
- Sequence:KQEYDEAGPSIVHRKC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping temperature:4 °C
- Immunogen:This ACTA1/Alpha-actin antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 346-375 amino acids from the C-terminal region of human ACTA1/Alpha-actin.
- Purification:Protein A & Peptide affinity purification
- Cat. no.:89512-280
- Supplier No.:AP14779B
Spécifications
A propos de cet article
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50, Flow Cytometry: 1:10~50, Immunofluorescence: 1:10~50,
Type: Primary
Antigen: ACTA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human