ContactUsLinkComponent
Polyster Foam Test Tube and Microcentrifuge Tube Racks, Bio Plas
Catalog # 60828-916
Supplier:  Bio Plas
undefined
Polyster Foam Test Tube and Microcentrifuge Tube Racks, Bio Plas
Catalog # 60828-916
Supplier:  Bio Plas
Supplier Number:  0010
Specifications
  • Color:
    Charcoal
  • Description:
    Fits 50 Tubes of sizes 10 x 75 mm, 12 x 75 mm, 13 x 100 mm
  • Cat. no.:
    60828-916
  • Supplier No.:
    0010
Specifications
About this item
Polyester foam racks securely hold and lock 50 culture tubes and 30 microcentrifuge and micro blood collecting tubes.
  • Flame retardant
  • Chemical and solvent resistant
  • Will not tip over easily
  • Weighs less than two ounces
Foam racks float on H2O bath with full tube and sample. Ideal for refrigerated or frozen storage samples. All tubes can be decanted simultaneously. For more uniform incubation, tubes may be pushed down into H2O bath while still being held securely in rack. Rack holds blood drawing tubes securely, and may therefore be used in blood drawing trays.