Borosilicate Glass Beads, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Solid borosilicate spheres.
- Beads have excellent chemical resistance
- Can be used as distillation column packing
- Can be used as boiling stones
- Can be used to provide support for chromatography columns used in gel filtrations
Packaging: Packed in boxes containing one pound.
Customers Who Bought This Also Bought