ContactUsLinkComponent
Fungi-Fluor® Pneumocystis Kit, Polysciences
Fungi-Fluor® Pneumocystis Kit, Polysciences
# de catalogue 10818-624
Fournisseur:  Polysciences Inc
Numéro CAS:  
Fungi Fluor* Pneumocystis kit
Fungi-Fluor® Pneumocystis Kit, Polysciences
# de catalogue 10818-624
Fournisseur:  Polysciences Inc
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Fungi-Fluor® Pneumocystis Kit
  • Cat. no.:
    10818-624
  • No. of Reactions:
    100
  • Supplier No.:
    22363
Spécifications
A propos de cet article
The Fungi Fluor® Pneumocystis Kit offers a fast, fluorescent staining procedure for Pneumocystis jiroveci in bronchial specimens.
Kit contains 10mL of Fungi-Fluor® staining solution, 10 positive unstained control slides, and Direct fluorescent stain for Pneumocystis carinii cyst.
Customers Who Bought This Also Bought