DuPont™ Proshield® 10 Lab Coats with Knit Collar and Cuffs
Fournisseur: DuPont
Certificats
A propos de cet article
DuPont™ ProShield® 10, spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont.
- Knit collar
- Knit cuffs
- Set sleeve design
- Length extends below hips
- Front snap closure
- Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments. DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Applications include general maintenance, janitorial/cleaning and other dirty work. Lab coats feature knit collar and cuffs, with a set sleeve design.
Certifications: Thickness meets ASTM D1117.
Spécifications
- Catalog No:
- 10016-294
- 10016-296
- 10016-306
- 10016-308
- 10016-310
- 10016-312
- 10016-290
- 10016-298
- 10016-300
- 10016-302
- 10016-304
- 10016-292
- 10016-314
- 10016-316
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
Spécifications
Informations connexes