About this item
Rxi® fused silica capillary GC columns are designed specifically to deliver exceptional inertness, ultra-low bleed, and reproducible retention and selectivity.
- General-purpose columns exhibiting extremely low bleed
- Ideal for many GC-MS applications, including pesticides, PCB congeners (e.g., Aroclor mixes), PAHs
- Unique selectivity
- Temperature range: 30 to 360 °C
- Low-polarity proprietary phase
This combination of column characteristics assures Rxi® columns will have long lifetimes.
Improvements in polymer synthesis and tubing deactivation enable inert, stable Rxi®-XLB columns especially well-suited for analyzing active, high molecular weight compounds with sensitive GC-MS systems, including ion trap detectors. Excellent efficiency, coupled with inertness, low bleed, and high thermal stability, make Rxi®-XLB columns ideal for analyzing semi-volatile compounds in drinking water.
Restek has also developed a stable, reproducible manufacturing process, so that Rxi® columns will meet your needs every time you install a new column.
Specifications
- Catalog No:
- 10058-454
- 10058-456
- 10058-450
- 10058-452
- 10058-440
- 10058-442
- 10058-438
- 10058-448
- 10058-444
- 10058-446
- 10850-546
- 10850-472
- 10059-550
- Description:
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Rxi-XLB
- Temperature limit:
- 30 to 340/360 °C
- 30 to 340/360 °C
- 30 to 340/360 °C
- 30 to 340/360 °C
- 30 to 340/360 °C
- 30 to 340/360 °C
- 30 to 340/360 °C
- 30 to 340/360 °C
- 30 to 340/360 °C
- 30 to 340/360 °C
- 30 to 340/360 °C
- 30 to 320/360 °C
- 30 to 340/360 °C