Plastic Disposable Syringes, SUN SRi
Supplier: Thermo Scientific
Certificates
About this item
Disposable syringes made of inert polypropylene and polyethylene.
- Chemically resistant
- Easy to read permanent graduations
- Safe functional back-stop feature
No rubber plunger seals or silicone lubricants that may cause sample contamination.
Packaging: Bulk packaged.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.