Pressure Refill Reaction Vials, Chemglass
Supplier: Chemglass
Certificates
About this item
Made from 33 expansion borosilicate glass, ideal for use in heated reactions.
- High resistance to thermal shock
- Maximum chemical resistance
Certifications: Conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585.
Delivery: Supplied with a special open top cap which prevents the pressure in the vial from exceeding 150 psig.
Packaging: Packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification.
Customers Who Bought This Also Bought