Sealing Films, Alumaseal 96, Chemglass
# de catalogue 30623-776
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Length:4 3/5 "
- Sterility:Non sterile
- Width:3"
- Description:Sealing film, AlumaSeal 96
- Thickness:1.5 mil
- Supplier No.:CLS-4001-009
Spécifications
A propos de cet article
Sealing film made of thick aluminium is pierceable and ideal for use with 96-well plates.
- Sealing films have one partial width end tab with no perforations
- Fits inside the rim of raised rim plates
- DNase, RNase and Nucleic acid free
Customers Who Bought This Also Bought