A propos de cet article
New globally harmonized chemical labeling rules have been enacted to protect workers who come into contact with hazardous chemicals.
- Bottles are made from high-purity FEP for low extractables and long life in the lab, and are suitable for dispensing high-purity solvents
- Permanently laser etched with 100% Globally Harmonized System (GHS) chemical safety information
- No other system markings are used on the bottles, which eliminates confusion
- Safety information is laser etched directly into the wall of the bottle so information will remain permanent and legible for the life of the wash bottle
- Dispensing nozzles are carefully molded to deliver a straight stream for precision dispensing and safety
- Designed to dispense through the top with a closure and stem molded in one piece for uniform, leakproof service
Nalgene Right-to-Understand FEP Wash Bottles are ready to use when transfering common laboratory chemicals from manufacturers’ containers. They are pre-labeled with chemical identification information, hazard and precautionary statements, GHS pictograms, CAS, and EC numbers. Wash bottles are available for use with acetone, toluene, n-hexane, methylene chloride, xylene, and methyl ethyl ketone (MEK).
Primary chemical containers are now labeled with new Globally Harmonized System icons and cautions. To avoid confusion and protect employees, secondary containers like wash bottles should have the same label information. Using Nalgene® Right-to-Understand Wash Bottles takes the guesswork out of chemical safety labeling, and helps prevent chemical related accidents and illnesses in the lab.