ContactUsLinkComponent
Disposable Medicine Cups, Plastic, Sklar® Premium
Disposable Medicine Cups, Plastic, Sklar® Premium
# de catalogue 10193-848
Fournisseur:  Sklar
Medicine Cup Sterile 2 oz, Plastic, Disposable, Latex-Free, Premium OR Grade
Disposable Medicine Cups, Plastic, Sklar® Premium
# de catalogue 10193-848
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Capacity:
    59 ml (2 oz.)
  • Description:
    Medicine Cups
  • Supplier No.:
    96-7027
Spécifications
A propos de cet article
Sterile, disposable transparent plastic sample cups for medicinal purposes.
  • Latex free
  • Graduated
  • Premium OR-Grade
Sklar's® Medicine Cup is used for dispensing both liquid and dry medications. The cups are made of polypropylene and are latex-free. They have easy to read markings on the interior surface
Customers Who Bought This Also Bought