Super Nuova+™ Hot Plate Stirrer, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Thermo Scientific™ SuperNuova+™ stirring hotplate performs with dependable safety and simple operation, units are available in two sizes including a multi-position model.
- Top plate choices in ceramic or aluminum
- Easy-to-read LED display for heating and stirring
- Raised display design protects electronics from spills
- Ability to set programs (ramp/dwell) as well as save settings
- Optional PT1000 temperature probe and splash guard protection shield
Stir trac features for slow-speed stirring, consistent speed control, and stronger magnetic coupling. Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F).
Customers Who Bought This Also Bought