Spécifications
- Length:8.2 cm (31/4")
- Sterility:Non sterile
- Description:Ives Rectal Speculum with Hard Rubber Obturator
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Material:Rubber
- Supplier No.:80-2245
Spécifications
A propos de cet article
Sklar® Ives Rectal Speculum, also known as the Ives-Fansler Anoscope, is used for examining the anal canal, hemorrhoidectomy, and rectal excision.
Non-latex, plastic obturator used to assist with insertion of the speculum. Diameter at the tip is 18 mm and diameter nearest the handle is 25 mm.