Hydrion™, Mikro pH Test Paper with Dispensor, Micro Essential Laboratory
# de catalogue 60775-881
Fournisseur: Micro Essential
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:pH Paper, Mikro Hydrion Dispenser Double Roll (1-12)
- Cat. no.:60775-881
- Supplier No.:M-1200
Spécifications
A propos de cet article
Hydrion® Double Roll pH Paper set the standard for quality, accuracy, and reliability
- Composed of Special Wet Strength Paper
- Color chart provides clear color matches based on spectral color changes for sample comparison
- These Mikro paper rolls measure 180 x 5/32 inches
These rolls test the 1.0-12.0 pH range with each full pH unit showing a distinct color variation.
Packaging: Packaged in a dual roll protective plastic dispenser and contained in a polypropylene case to guard against sunlight and laboratory hazards.
Customers Who Bought This Also Bought