PYREX® Folin-Wu Digestion Tube, Corning
# de catalogue 26658-040
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Volume:50 ml
- Description:PYREX® Folin-Wu Digestion Tube
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- O.D.×L:25×200 mm
- Tolerance:±0.4 ml
- Supplier No.:7900-25
Spécifications
A propos de cet article
Made of PYREX® borosilicate glass.
- Round bottom
- With engraved white-filled graduations at 25 and 50 ml
- Tolerance of ±0.4 ml
The Folin-Wu digestion tubes are graduated with a beaded rim and are fabricated from specially selected tubing for product durability.
Customers Who Bought This Also Bought