Spécifications
- Length:16.5 cm (6.5")
- Description:Murphy Plaster Knife, Premium OR-Grade
- Tip Configuration:Blunt
- Curvature:Curved
- Cat. no.:10194-972
- Handle Material:Wood
- Supplier No.:11-1540
- Blade Material:Stainless steel
Spécifications
A propos de cet article
Murphy Plaster Knife features a wide handle and a wide, slightly curved blade used for removing pieces of a dried plaster cast
- Made up of stainless steel
Censitrac Ready products are industry standard.
Instrumentation used for casting is typically stronger and heavier than standard instruments in order to adequately handle the heavy cast material. This plaster knife is equipped with a wooden handle.