ESD Inspection Gloves, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
A propos de cet article
Transforming Technologies ESD inspection gloves are designed to be worn when handling ESD susceptible items.
- Breathable
- Laundered for repeated use
Gloves are manufactured from a nylon and conductive carbon fabric, and feature finished rolled cuffs. Tip or palm coatings consist of micro-foamed polyurethane coating.
They are made with a dissipative nylon to reduce charge generation. A seamless fine gauge knit construction makes it the most breathable glove available. Available in plain, polyurethane finger tip coated, or polyurethane palm coated formats.
Certifications: Inspection gloves offer a resistance of 10⁶ - 10⁸ per ANSI/ESD SP15.1.
Packaging: Sold in packs of 12, one size per pack.
Spécifications
- Catalog No:
- 10809-008
- 10809-010
- 10809-004
- 10809-026
- 10809-006
- 10809-000
- 10809-022
- 10809-024
- 10809-002
- 10809-018
- 10808-996
- 10809-020
- 10808-998
- 10809-014
- 10808-992
- 10809-016
- 10808-994
- 10809-012
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable