ContactUsLinkComponent
Rainbow Transformation Kit
Rainbow Transformation Kit
# de catalogue 470136-456
Fournisseur:  Edvotek
Rainbow Transformation kit
Rainbow Transformation Kit
# de catalogue 470136-456
Fournisseur:  Edvotek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Rainbow transformation kit
  • No. of Lab Groups:
    10
  • Cat. no.:
    470136-456
  • Supplier No.:
    224
Spécifications
A propos de cet article
In this colorful experiment, students will explore the biological process of bacterial transformation using vibrant chromogenic proteins.
  • Explore genetic engineering and the central dogma with this colorful experiment
  • Transform E.coli with plasmids that contain a gene for ampicillin resistance and a gene for either a blue, purple, or pink pigment
  • Select for transformed cells using LB-ampicillin plates and calculate transformation efficiency
  • Expose transformed cells to IPTG to demonstrate differential gene expression
  • Different plasmids help emphasize the concept of DNA>RNA>Protein>Trait
Students will use three recombinant plasmids to transform E.coli bacteria. The resulting E.coli will be examined for the presence of blue, purple, and pink pigments, as well as for resistance to ampicillin. Features our new enhanced transformation protocol for improved student results.
Ordering information: Note: If shipping to warehouse, perishable coupon included.
Customers Who Bought This Also Bought