MyCapLC™ Kits, Trajan Scientific and Medical
Fournisseur: Trajan Scientific and Medical
Certificats
A propos de cet article
Customize CapLC for your application using the empty hardware provided.
- DIY capillary column packing, no special intrumentation required
- Make the columns that you want
- Perfect for your life sciences analysis
- Use the C18 media provided or use your own media
MyCapLC Kit enables you to create your own CapLC for higher resolution analysis compared to traditional HPLC. Includes simple packing instructions and you can easily pack your own capillary columns with speed in a matter of minutes using your own HPLC pump.
Capillary Liquid Chromatography (CapLC) is a common technique in the life sciences, where sample amounts are limited and the analyte of interest might be of very low concentration. MyCapLC™ Kit enables you to create your own CapLC for a higher resolution analysis compared to traditional HPLC.
Certifications: RoHS compliant per Directive 2011/65/EU.
Ordering information: MyCapLC Kit contains everything you need to pack three columns, including 0.1 g C18 3 μm 200 Å, 1 slurry reservoir (50×2.1 mm Int.Ø column tubing), 2 end fittings, 2 PEEK seals, 1 PEEK Fingertight fitting, 3 column bodies (¹/₁₆" Ext.Ø PEEKsil, 100 x0.3 mm Int.Ø), 6 connection capillaries (¹/₃₂" Ext.Ø, 100 mm × 50 µm Int.Ø), 6 male unions (¹/₁₆"), 6 female unions (¹/₃₂"), 6 PEEK double ended ferrules (¹/₁₆ to ¹/₃₂"), 6 woven mesh filters (0.5 µm, ¹/₁₆") and 2 wrenches.
Customers Who Bought This Also Bought