Chemotherapy Waste/Transport Bags, Unimed
Fournisseur: Unimed-Midwest
Certificats
A propos de cet article
Extra-thick, large capacity polyethylene bags provide extra protection in chemotherapy waste handling and transport.
- All bags are CYTA labeled
The 22.7 L (6 gal.) bag is equipped with a convenient and safe zip closure.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.