Raptor™ EXP® Guard Column Cartridges, Restek
Fournisseur: Restek
Certificats
A propos de cet article
To help protect your investment and further extend the life of Restek's already-rugged LC columns, Restek offers the patent-pending guard column hardware developed by Optimize Technologies.
- Available in a number of sizes
- Maximum cartridge pressure: 600 bar/8700 psi (2.7 µm) or 400 bar/5800 psi (5 µm)
A Restek LC guard cartridge in an EXP® direct connect holder is the ultimate in column protection.
Customers Who Bought This Also Bought