A propos de cet article
The Ultra Carbamate column was designed specifically for carbamates analysis.
- Particle size: 3 µm or 5 µm, spherical
- Pore size: 100 Å
- pH range: 2.5 to 8
- Temperature limit: 80 °C
The unique packing separates 10 target carbamates in just over 10 minutes.
For post-column derivatization/fluorescence detection applications using a 4.6 mm Int.Ø column, the total system dead volume, including the post-column reactor, must be less than 650 µl. For standard post-column reactor systems, we recommend a 250×4.6 mm, 5 µm column. Contact Restek Technical Service or your local Restek representative for more information.
The column is compatible with fluorescence or LC-MS detection. An Ultra Carbamate column can process as many as three samples per hour, versus less than two samples per hour on a general-purpose C18 column. In addition to increased sample throughput, this much faster analysis will significantly reduce solvent usage—and the costs of disposing of solvent waste.