ContactUsLinkComponent
Ward’s® Transformation of E.coli: Ampicillin Resistance
Ward’s® Transformation of E.coli: Ampicillin Resistance
Catalog # 470007-116
Supplier:  Avantor
undefined
Ward’s® Transformation of E.coli: Ampicillin Resistance
Catalog # 470007-116
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Transformation of E.coli: Ampicillin Resistance Kit
  • No. of Lab Groups:
    6
  • Cat. no.:
    470007-116
Specifications
About this item
What is genetic engineering?
  • Includes pre-poured petri plates
  • Links to real world concepts
  • Estimated class/lab time required: 45 minutes over 2 days
Transformation, a fundamental technique of genetic engineering, involves the induced uptake of foreign DNA by a host cell. Based on the gene(s) encoded by the introduced DNA, transformation may confer a new trait or traits to the host cell. Using a plasmid which carries a gene for antibiotic resistance to ampicillin, students investigate one mechanism of bacterial transformation into wild-type E.coli cells. This lab activity also may be extended into discussions of the growing threat of antibiotic-resistant strains of pathogenic bacteria. The reliable and convenient lab activity that can be performed in one class period without the use of a centrifuge. For your convenience, pre-poured cultured plates are provided with culture material. This activity is similar to the classic AP 6A Lab.
Ordering information: A coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.
Frequently Bought Together