ContactUsLinkComponent
Superior Jolly Bulb Metal
Superior Jolly Bulb Metal
# de catalogue 470157-716
Fournisseur:  Wards
Jolly Bulb Metal
Superior Jolly Bulb Metal
# de catalogue 470157-716
Fournisseur:  Wards
Spécifications
  • Description:
    Jolly Bulb Metal, Superior
  • Cat. no.:
    470157-716
  • Measurement Range:
    –1 to 32 Kg/cm, Manometer
  • Supplier No.:
    PH0138D
Spécifications
A propos de cet article
Visually impacting demonstration of the relationship between temperature and pressure.
  • Easy to read pressure gauge
  • Instruction manual
  • Metal sphere to insure constant volume
A hollow metallic ball 80 mm in diameter is connected to an absolute pressure gauge. Submerging the metal ball in ice water causes pressure to drop, while submerging it in hot water increased the pressure. Utilizing several data points students can calculate the value for absolute zero.
The Superior Jolly Bulb has two scales. The inside scale measures in kPa and the outside scale measures in PSI.
Customers Who Bought This Also Bought