Accessories for Gas Sampling Valves and Replacement Rotors (Sulfinert®-Treated), Restek
Fournisseur: Restek
Certificats
A propos de cet article
Replacement rotors are available for use with 4-, 6-, and 10-port Sulfinert gas sampling valves.
- Rotors can be matched with different gas sampling valves depending on number of ports
Customers Who Bought This Also Bought