ALTEF® Gas Sampling Bags, Restek
# de catalogue 10803-304
Fournisseur: Restek
Spécifications
- Capacity:1 L
- W×L:7×7"
- Description:ALTEF gas sampling bag with PP combo valve and septum
- Cat. no.:10803-304
- Supplier No.:22959
Spécifications
A propos de cet article
Sampling bags are a low-cost whole air sampling device for high level VOCs and permanent gases.
- Excellent low-cost alternative to Tedlar® bags for collection of most VOCs
- Very low VOC and sulfur background compared to Tedlar® bags
- Not recommended for ketones, acetates, hydrogen sulfide, or permanent gases
- Durable PVDF film is resistant to abrasion and chemicals, including most acids and organic compounds
- Contain no additives, fillers, or pigments
Several EPA, NIOSH, and OSHA methods exist for bag sampling for a variety of applications: stationary sources emissions, workplace atmospheres, ambient, indoor air quality, and breath analysis. Choose the film type appropriate for the application. All Restek's bags feature a polypropylene combo valve with hose connection to fit ³/₁₆" Int. Ø tubing and syringe port with replaceable septum. A single eyelet provides handling convenience.
Customers Who Bought This Also Bought