ContactUsLinkComponent
Cell Organizer for ASE® Parts, Restek
Cell Organizer for ASE® Parts, Restek
# de catalogue 10015-356
Fournisseur:  Avantor
Cell Organizer for ASE Parts, blue 13-bin unit, 12in l x 12in h x 7.5in d
Cell Organizer for ASE® Parts, Restek
# de catalogue 10015-356
Fournisseur:  Avantor
Spécifications
  • Color:
    Blue
  • Description:
    Cell Organizer for ASE Parts, 13-Bin Unit
  • Cat. no.:
    10015-356
  • W×D×H:
    12×7.5×12"
  • Supplier No.:
    23998
Spécifications
A propos de cet article
Organizers are designed for use with ASE® parts.
  • Convenient storage of extraction cell parts and consumables
  • Thirteen open bins provide easy visibility and organization for small and large pieces
  • Small footprint conserves valuable lab bench and drawer space
Customers Who Bought This Also Bought