Replacement Swagelok® SS4H Bellows Valve, Restek
# de catalogue 10145-862
Fournisseur: Restek
Spécifications
- Description:Replacement 1/4" Swagelok SS4H Bellows-Sealed Valve (2-port),
- Material:Stainless Steel
- Cat. no.:10145-862
- Supplier No.:24148
Spécifications
A propos de cet article
Swagelok® SS4H bellows valves are constructed entirely of metal to prevent sample adsorption and deliver optimal accuracy with results.
- All-metal flow path prevents sample adsorption, giving more accurate results
- Unique serial number on each valve for complete traceability
- Withstands temperatures of up to 300°C
- Rugged performance in the field
Products are completely traceable with unique serial numbers on every unit. Durable fabrication ensures rugged performance in the field.
Ordering information: Replacement 1/4" Swagelok SS4H bellows-sealed valves are available on SilcoCan canisters as a custom product.
Customers Who Bought This Also Bought