Sample Collection Bottles for ASE® 100/300 Systems, 250 ml, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Cleaned, ready-to-use sample collection bottles are suitable for use with ASE® 100/300 systems.
- Cleaned, assembled, and ready to use
- Collection bottles with volume of 250 mL
- Clear or amber glass
- Caps and PTFE-lined septa included
For use with ASE® 100/300 systems.
Customers Who Bought This Also Bought