Extraction Cell Bodies for ASE® Systems, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Accelerated solvent extraction is a common technique for fast and reliable extraction of organic materials from solid matrices, using EPA SW-846 Method 3545, Pressurized Fluid Extraction (PFE).
- Cell bodies are serialized for easy sample identification
- Smooth inner surfaces for easier cleaning
Restek offers a wide range of replacement parts to keep your extraction system running smoothly. All parts are economically priced to save you money and are designed to meet or exceed the performance of the original manufacturer’s parts.
Customers Who Bought This Also Bought