Spécifications
- Cat. no.:10015-398
- Supplier No.:26033
Spécifications
A propos de cet article
Diatomaceous earth is used as a filter aid to improve extraction efficiency of densely packed soils, such as clays.
- Improves extraction efficiency
- Adsorbs moisture from samples
By mixing the sample with diatomaceous earth, recoveries can be improved and excess moisture can be absorbed. Packaged in a convenient 1 kg quantity.