Sample Cylinders, High Pressure (Stainless Steel & Sulfinert®-Treated), Restek
Fournisseur: Restek
Certificats
A propos de cet article
Swagelok® sample cylinders are made of 304L and 316L stainless steel to resist corrosion and are DOT rated to 1,800 and 5,000 psig (TPED cylinders rated to 1,450 and 4,350 psig), which allows sampling at gas wellheads as well as on-site refineries.
- 304L stainless steel; DOT rating to 1,800 psig (TPED cylinders to 1,450 psig)
- Range of cylinder sizes, from 75 cc to 2,250 cc
- All cylinders have 1/4" female NPT threads on both ends
- TPED compliant cylinders available for EU community
Each cylinder is hydrostatically tested to at least 5/3 the working pressure.
Customers Who Bought This Also Bought