Science Take-Out® Gene Switches
Fournisseur: SCIENCE TAKE-OUT, LLC
Certificats
A propos de cet article
How Do Bacteria Turn On And Turn Off Genes?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
Switching genes on and off enables bacteria to conserve resources by producing proteins only when they are needed. Make a model of an operon. Learn how the lac operon regulates the expression of bacterial genes essential for lactose metabolism. This complete "dry lab" activity contains all required materials.
Customers Who Bought This Also Bought