ContactUsLinkComponent
Orion Dermatoscope, OR Grade, Sklar
Orion Dermatoscope, OR Grade, Sklar
# de catalogue 10194-704
Fournisseur:  Sklar
Dermatoscope
Orion Dermatoscope, OR Grade, Sklar
# de catalogue 10194-704
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Orion Dermatoscope
  • Cat. no.:
    10194-704
  • Supplier No.:
    06-6250
Spécifications
A propos de cet article
Sklar's Orion Dermatoscope is a dermal diagnostic instrument used in the examination of skin lesions
  • Stainless Steel/Plastic
It is an oil immersion/non-polarized instrument that utilizes 10X magnification and illumination to closely examine skin lesions, melanoma, and infections. Oil or petroleum jelly may be used with the graduated plate to provide additional contrast and definition. The handle of the Orion Dermatoscope features rheostats that adjust the light intensity with a simple rotation.
Customers Who Bought This Also Bought