Somso® Male Torso with Head and Open Back
# de catalogue 470165-202
Fournisseur: MARCUS SOMMER
Spécifications
- Description:Male Torso with Head and Open Back
- Cat. no.:470165-202
- Supplier No.:AS23/1
Spécifications
A propos de cet article
This is natural size, in SOMSO-Plast®.
Separates into 20 parts: brain with arteries (4), eye with muscles and optic nerve, halves of the lung (2), heart (2), liver, stomach (2), small and large intestine (3), opening appendix, omentum, half of right kidney, half of bladder, torso. On a green base. Height: 86 cm.
Customers Who Bought This Also Bought