ContactUsLinkComponent
Silicone Heat Transfer Fluid 510, FoxWest Sales
  10611-548
 :  
undefined
Silicone Heat Transfer Fluid 510, FoxWest Sales
  10611-548
 :  2101
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Silicone heat transfer fluid 510, 3.8 L (1 gal.)
  • Cat. no.:
    10611-548
  • Supplier No.:
    2101
Silicon Heat Transfer Fluid 510 is a methyl-phenyl polysiloxane fluid designed for applications that require ultra-violet light protection or applications that require thermal stability at high temperatures
The working temperature range for silicone bath fluid is 0 to 88°C (32 to 190°F). The fluid viscosity is 14cs.