Via Blood Specimen Transporter Tote, Sonoco ThermoSafe
Fournisseur: Sonoco Thermosafe
Certificats
A propos de cet article
Insulated expanded polypropylene medical transporter tote is an insulated carrier ideal for on-site collection, storage and transport of temperature-sensitive specimens, medications, medical devices, and organs.
- Ideal for storage of blood products
- ZERO BENCH-TIME™ refrigerant technology
- Pre-qualified to maintain packed red blood cells
Transporters are ideal for the temporary storage of blood and blood products in the operating room and the transport of medications, medical devices, specimens and organs
Simple, reusable ergonomic design utilizes leading edge technology to provide ultimate temperature protection.
ZERO BENCH-TIME™ refrigerant technology reduces assembly process for time and temperature critical materials. Pre-qualified to maintain packed red blood cells at 1-6°C and 1-10°C.
Customers Who Bought This Also Bought