ContactUsLinkComponent
Bluing Tablets, FoxWest Sales
Bluing Tablets, FoxWest Sales
# de catalogue 10611-544
Fournisseur:  Decon Labs
Tablets bluing
Bluing Tablets, FoxWest Sales
# de catalogue 10611-544
Fournisseur:  Decon Labs
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Bluing Tablets
  • Cat. no.:
    10611-544
  • Supplier No.:
    1220
Spécifications
A propos de cet article
Bluing Tablets are used to color the water in a toilet, via a blue dye, in order to prevent the water from being used to dilute a urine sample for drug testing
Packaged two tablets per foil packet, bluing tablets are an efficient and inexpensive means of preventing tampering to critical drug sampling.
Customers Who Bought This Also Bought