ThermoSafe® LabWare Low-Temp Chiller Insulated Shippers, Sonoco ThermoSafe
Fournisseur: Sonoco Thermosafe
Certificats
A propos de cet article
ThermoSafe® LabWare foam shippers are versatile insulated shippers and aid temperature-sensitve enzymes and reagents at workstations.
- Lightweight
- Easy-to-handle
- Protects temperature-sensitive enzymes and reagents
- Low-temperature baths
Durable components are designed for convenience while in transit or in-use
The Low-Temp Chiller is an economical cold trap or alcohol/dry ice bath. Convenient shallow depth has 38 mm (1 ½") insulating walls.
Holds wet or dry ice, alcohol.
These shippers are lightweight and easy to handle.
Customers Who Bought This Also Bought