Spécifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:White
- Glove size:L
- Description:BCR® Polyester Full-Finger Glove Liner
- Cat. no.:89233-406
- Supplier No.:BGL3.8LB
Spécifications
A propos de cet article
BCR full finger polyester glove liners, large (blue cuff).
- Full line offers a choice to better provide even the most discriminating wearer with the most appropriate glove liner
- Due to specific engineering, BCR® Glove Liners fit most hands up to a size 9
- This allows for easier stock control, while the availability of a larger size accommodates the hard to fit hands
Composition and attributes: 100% continuous filament, knitted polyester. Composition allows moisture to be absorbed and wicked away from skin during extended wear. Constructed using special seamless techniques. No-run and non-ravel. Ambidextrous for easy stock control. Launderable for multiple reuse. The glove liner is suitable in any ISO class cleanroom when worn under a nitrile, latex, vinyl, neoprene or other type glove.
BCR® Polyester Glove Liners - Protection and comfort during extended wear. BCR® Glove Liners are the choice in any industry, when the job calls for long hours of use when hand comfort is a concern. Special fiber construction allows for moisture absorption during extended wear.