Banana to alligator clip wires
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Easily connect power supplies to devices without electrical posts.
- One Alligator Clip
- One Stackable Banana Clip
- Banana Clip Size: 4 mm
- Various Lengths Available
- Flexible, 18-Gauge Stranded Wire Cord
A flexible, 18-gauge stranded wire cord has a stackable, 4 mm banana clip at one end and an alligator clip at the other.
Alligator clips have a protective vinyl sheath. They're available in various lengths and pack sizes.
Customers Who Bought This Also Bought