BCR® Polyester Cleanroom Glove Liners, Berkshire
# de catalogue 89212-859
Fournisseur: Berkshire
Small Business Enterprise
Spécifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:White
- Glove size:L
- Description:Half-finger gloves
- Disposable / reusable:Reusable
- Cat. no.:89212-859
- Packaging:Bulk, 200 Pairs per pack
- Supplier No.:BGL2.8LB
Spécifications
A propos de cet article
BCR® cleanroom glove liners enhance operator comfort.
- 100% continuous filament knitted polyester
- Polyester offers a thinner yet durable barrier for the wearer
- Half-fingered design covers palm and parts of fingers to permit tactile use while wicking moisture from palms
- Reusable, ambidextrous, seamless construction
For use in laboratories and manufacturing environments. Can be used alone as inspection gloves to prevent fingerprints on circuit boards, glassware, lenses, optics and other critical surface.
Suitable in any ISO class cleanroom when worn under a nitrile, latex, vinyl, neoprene or other type glove.
Customers Who Bought This Also Bought