ContactUsLinkComponent
Ecobond® Cleanroom Paper, Berkshire
Catalogue # 89097-760
Fournisseur:  Berkshire
Small Business Enterprise
   Waste Reducing
undefined
Ecobond® Cleanroom Paper, Berkshire
Catalogue # 89097-760
Fournisseur:  Berkshire
Supplier Number:  ECO85.0811WH
Small Business Enterprise
   Waste Reducing
Spécifications
  • Color:
    White
  • Dimensions:
    8.5×11"
  • Environmentally Preferable:
    Waste Reducing
  • Cat. no.:
    89097-760
  • Weight:
    85.5 g/m² (24#)
  • Supplier No.:
    ECO85.0811WH
Spécifications
À propos de cet article
EcoBond® is a latex free recyclable version of BCR® cleanroom bond
  • Low extractables and ions
  • Compatible with toner and offset printing
  • High strength and heat resistance
  • Good toner adhesion
  • Cleanroom Class 1 - 100,000
EcoBond® designed for high level of contamination control in critical processing applications. Cellulose sourced from sustainable managed forests.
Compatible with standard copiers and printers. Not recommended for inkjet printers.
EcoBond® is made from pure and sustainable source of cellulose as standard BCR® cleanroom bond, and is recommended for use in ISO class 3 to 8 cleanrooms.
Certifications: Complies with RoHS and REACH requirements.
Ordering information: 250 sheets per pack, 10 packs per case (double packed).